accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSClOO

CAS RN 98-68-0

IUPAC Name: 4-methoxybenzene-1-sulfonyl chloride
Synonyms: 4-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 206.64
Molecular Formula: C7H7ClO3S
InChi Key: DTJVECUKADWGMO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)