accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOSClOO

CAS RN 98-74-8

IUPAC Name: 4-nitrobenzene-1-sulfonyl chloride
Synonyms: 4-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol): 221.61
Molecular Formula: C6H4ClNO4S
InChi Key: JXRGUPLJCCDGKG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)