accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98-78-2

IUPAC Name: 3-oxocyclopentane-1-carboxylic acid
Synonyms: 3-oxocyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: RDSNBKRWKBMPOP-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results