accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOOHN

CAS RN 98-79-3

IUPAC Name: 5-oxopyrrolidine-2-carboxylic acid
Synonyms: pyroglutamate acidum pidolicum pidolate, magnesium DL-pyroglutamic acid L-glutiminate acid, pyrrolidonecarboxylic glutiminate ajidew A-100 (S)-pyroglutamate glutimate
Molecular Weight (g/mol): 129.12
Molecular Formula: C5H7NO3
InChi Key: ODHCTXKNWHHXJC-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC(=O)N1
Molecular Weight (g/mol) 
  • (4)
  • (6)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results