accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98-82-8

IUPAC Name: (propan-2-yl)benzene
Synonyms: cumene isopropilbenzene cumene (substance) 2-fenilpropano rcra waste number U055 cumeen 2-fenyl-propaan isopropylbenzeen
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: RWGFKTVRMDUZSP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (9)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (7)
Percent Purity 
  • (2)
  • (5)
  • (3)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (9)
  • (6)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (7)
  • (2)
  • (5)
  • (3)
  • (10)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)