accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98-83-9

IUPAC Name: (prop-1-en-2-yl)benzene
Synonyms: α-methylstyrol as-methylphenylethylene 1-methylethenylbenzine 1-methylethylenebenzene isopropenyl-benzeen α-methylstyreen α-methyl-styrol α-metil-stirolo isopropenil-benzolo
Molecular Weight (g/mol): 118.18
Molecular Formula: C9H10
InChi Key: XYLMUPLGERFSHI-UHFFFAOYSA-N
SMILES: CC(=C)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results