accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONO

CAS RN 98-95-3

IUPAC Name: nitrobenzene
Synonyms: nitrobenzene oil of mirbane
Molecular Weight (g/mol): 123.11
Molecular Formula: C6H5NO2
InChi Key: LQNUZADURLCDLV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (14)
Percent Purity 
  • (5)
  • (3)
  • (5)
  • (4)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (8)
  • (3)
  • (1)
Physical Form 
  • (11)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)
  • (14)
  • (5)
  • (3)
  • (5)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (3)
  • (1)
  • (11)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)