accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3NOHOOFNNHCH3F

CAS RN 98079-52-8

IUPAC Name: hydrogen 1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid chloride
Synonyms: hydrogen lomefloxacin chloride
Molecular Weight (g/mol): 387.81
Molecular Formula: C17H20ClF2N3O3
InChi Key: KXEBLAPZMOQCKO-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCN1C=C(C(O)=O)C(=O)C2=CC(F)=C(N3CCNC(C)C3)C(F)=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results