accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98330-04-2

CH3NH3CH3CH3CF

CAS RN 98330-04-2

IUPAC Name: tetraethylazanium hydrate fluoride
Synonyms: tetraethylammonium hydrate fluoride
Molecular Weight (g/mol): 167.27
Molecular Formula: C8H22FNO
InChi Key: YEKUWOWHPVKTCQ-UHFFFAOYSA-M
SMILES: O.[F-].CC[N+](CC)(CC)CC
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)