accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98432-80-5

H3CCH3OPOBrOCH3H3C

CAS RN 98432-80-5

IUPAC Name: bis(propan-2-yl) (bromomethyl)phosphonate
Synonyms: diisopropyl bromomethylphosphonate
Molecular Weight (g/mol): 259.08
Molecular Formula: C7H16BrO3P
InChi Key: JNONGIUWXHOEEN-UHFFFAOYSA-N
SMILES: CC(C)OP(=O)(CBr)OC(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results