accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98437-24-2

HOBHOO

CAS RN 98437-24-2

IUPAC Name: (1-benzofuran-2-yl)boronic acid
Synonyms: 1-benzofuran-2-ylboronic acid
Molecular Weight (g/mol): 161.95
Molecular Formula: C8H7BO3
InChi Key: PKRRNTJIHGOMRC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=CC=CC=C2O1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)