accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONH2ClCl

CAS RN 98446-49-2

IUPAC Name: 2,4-dichloro-5-methoxyaniline
Synonyms: 2,4-dichloro-5-methoxyaniline
Molecular Weight (g/mol): 192.04
Molecular Formula: C7H7Cl2NO
InChi Key: AJROJTARXSATEB-UHFFFAOYSA-N
SMILES: COC1=CC(N)=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results