CAS RN 985-16-0
CAS RN 985-16-0
IUPAC Name:
sodium (2S,5R,6R)-6-(2-ethoxynaphthalene-1-amido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms:
nafcillin sodium nafcillin sodium anhydrous
Molecular Weight (g/mol):
436.46
Molecular Formula:
C21H21N2NaO5S
InChi Key:
AYAPZOUDXCDGIF-FRFVDRIFSA-M
SMILES:
[Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Nafcillin sodium salt
Thermo Scientific Chemicals Nafcillin sodium salt
CAS: 985-16-0 Molecular Formula: C21H21N2NaO5S Molecular Weight (g/mol): 436.46 MDL Number: MFCD00056863 InChI Key: AYAPZOUDXCDGIF-FRFVDRIFSA-M Synonym: 4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylicacid, 6-2-ethoxy-1-naphthalenyl carbonyl amino-3,3-dimethyl-7-oxo-, sodiumsalt 1:1 , 2s,5r,6r PubChem CID: 131675018 IUPAC Name: (2S,5R,6R)-6-[(2-ethoxynaphthalene-1-carbonyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium SMILES: [Na+].CCOC1=CC=C2C=CC=CC2=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O