accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 98556-31-1

IUPAC Name: 4-chloro-6-iodoquinazoline
Synonyms: 4-chloro-6-iodoquinazoline
Molecular Weight (g/mol): 290.49
Molecular Formula: C8H4ClIN2
InChi Key: BDAIUOPDSRAOKI-UHFFFAOYSA-N
SMILES: ClC1=NC=NC2=CC=C(I)C=C12
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results