CAS RN 98737-29-2
(2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%
CAS: 98737-29-2 Molecular Formula: C15H21NO3 Molecular Weight (g/mol): 263.34 MDL Number: MFCD02258997 InChI Key: NVPOUMXZERMIJK-QWHCGFSZSA-N Synonym: 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane,2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane,tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate,2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane,tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate,tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate,tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate,2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane,ksc523o6b PubChem CID: 9903372 IUPAC Name: tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2
tert-Butyl N-[(1S)-1-((2S)-Oxiran-2-yl)-2-phenylethyl]carbamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S,3S)-3-Boc-amino-1,2-epoxy-4-phenylbutane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
(2S,3S)-3-Boc-amino-1,2-epoxy-4-phenylbutane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
(2S,3S)-3-Boc-amino-1,2-epoxy-4-phenylbutane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
(2S,3S)-3-(Boc-amino)-1,2-epoxy-4-phenylbutane, 98%, Thermo Scientific™
CAS: 98737-29-2 Molecular Formula: C15H21NO3 Molecular Weight (g/mol): 263.337 MDL Number: MFCD02258997 InChI Key: NVPOUMXZERMIJK-QWHCGFSZSA-N Synonym: 2s,3s-1,2-epoxy-3-boc-amino-4-phenylbutane,2s,3s-n-t-boc-3-amino-1,2-epoxy-4-phenylbutane,tert-butyl s-1-s-oxiran-2-yl-2-phenylethyl carbamate,2s,3s-1,2-epoxy-3-tert-butoxycarbonylamino-4-phenylbutane,tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenylethyl carbamate,tert-butyl s-r*,r*---1-oxiranyl-2-phenylethyl carbamate,tert-butyl n-1s-1-2s-oxiran-2-yl-2-phenyl-ethyl carbamate,2s,3s---3-t-boc-amino-1,2-epoxy-4-phenylbutane,ksc523o6b PubChem CID: 9903372 IUPAC Name: tert-butyl N-[(1S)-1-[(2S)-oxiran-2-yl]-2-phenylethyl]carbamate SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C2CO2