accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3COOHNO

CAS RN 98760-08-8

IUPAC Name: tert-butyl N-[1-(oxiran-2-yl)-2-phenylethyl]carbamate
Synonyms: tert-butyl N-[1-(oxiran-2-yl)-2-phenylethyl]carbamate
Molecular Weight (g/mol): 263.34
Molecular Formula: C15H21NO3
InChi Key: NVPOUMXZERMIJK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results