accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

S∗∗Men

CAS RN 98837-51-5

IUPAC Name: 3-butyl-2,5-dimethylthiophene
Synonyms: 3-butyl-2,5-dimethylthiophene
Molecular Weight (g/mol): 168.3
Molecular Formula: C10H16S
InChi Key: DUOSBQJOYVIVOR-UHFFFAOYSA-N
SMILES: CCCCC1=C(C)SC(C)=C1
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)