CAS RN 98892-75-2
IUPAC Name:
3-butyl-1,2-dimethyl-1H-imidazol-3-ium
Synonyms:
1-butyl-2,3-dimethylimidazol-1-ium
Molecular Weight (g/mol):
153.25
Molecular Formula:
C9H17N2
InChi Key:
XUAXVBUVQVRIIQ-UHFFFAOYSA-N
SMILES:
CCCC[N+]1=C(C)N(C)C=C1
1
–
1
of
1
results
1
1-Butyl-2,3-dimethylimidazolium chloride, 99%
CAS: 98892-75-2 Molecular Formula: C9H17ClN2 Molecular Weight (g/mol): 188.699 MDL Number: MFCD03093291 InChI Key: HHHYPTORQNESCU-UHFFFAOYSA-M Synonym: 1-butyl-2,3-dimethylimidazolium chloride,1-butyl-2,3-dimethyl-1h-imidazol-3-ium chloride,1-butyl-2,3-dimethylimidazol-1-ium chloride,3-butyl-1,2-dimethylimidazol-1-ium chloride,acmc-209sba,dsstox_cid_27882,dsstox_rid_82635,dsstox_gsid_47906,1-butyl-2,3-dimethylimidazoliumchloride,1-butyl-2,3-dime-thylimidazolium chloride PubChem CID: 2734167 IUPAC Name: 1-butyl-2,3-dimethylimidazol-3-ium;chloride SMILES: CCCCN1C=C[N+](=C1C)C.[Cl-]