accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H3CONHClClCl

CAS RN 98946-18-0

IUPAC Name: tert-butyl 2,2,2-trichloroethanimidate
Synonyms: tert-butyl 2,2,2-trichloroethanimidate
Molecular Weight (g/mol): 218.5
Molecular Formula: C6H10Cl3NO
InChi Key: CQXDYHPBXDZWBA-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=N)C(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)