accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHO

CAS RN 99-04-7

IUPAC Name: 3-methylbenzoic acid
Synonyms: toluic acid cadmium salt 3-toluic acid β-bethylbenzoic acid β-bethylbenzoate zinc salt 3-toluic acid barium salt 3-toluic acid β-methylbenzoic acid M-toluic acid
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: GPSDUZXPYCFOSQ-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)