accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OHO

CAS RN 99-05-8

IUPAC Name: 3-aminobenzoic acid
Synonyms: gabaculine 3-aminobenzoesaeure m-aminobenzoesaeure
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: XFDUHJPVQKIXHO-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (7)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)