CAS RN 99-19-4
CAS RN 99-19-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Prunasin, TRC
CAS: 99-19-4 Molecular Formula: C14H17NO6 Molecular Weight (g/mol): 295.29 Synonym: (αS)-α-(β-D-Glucopyranosyloxy)benzeneacetonitrile,L-Prunasin,Sambunigrin,(2S)-Sambunigrin; IUPAC Name: (2S)-2-phenyl-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyacetonitrile SMILES: C1=CC=C(C=C1)[C@@H](C#N)O[C@H]2[C@@H]([C@H]([C@@H]([C@H](O2)CO)O)O)O