accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99-20-7

IUPAC Name: 2-(hydroxymethyl)-6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol
Synonyms: trehalose
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: HDTRYLNUVZCQOY-UHFFFAOYNA-N
SMILES: OCC1OC(OC2OC(CO)C(O)C(O)C2O)C(O)C(O)C1O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)