accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOHHO

CAS RN 99-50-3

IUPAC Name: 3,4-dihydroxybenzoic acid
Synonyms: 3,4-dihydroxybenzoic acid protocatechuic acid protocatehuic acid
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: YQUVCSBJEUQKSH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)