accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99-51-4

CH3H3CNOO

CAS RN 99-51-4

IUPAC Name: 1,2-dimethyl-4-nitrobenzene
Synonyms: 1,2-dimethyl-4-nitrobenzene
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: HFZKOYWDLDYELC-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results