accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2H2NNOO

CAS RN 99-56-9

IUPAC Name: 4-nitrobenzene-1,2-diamine
Synonyms: 2-amino-4-nitroaniline 4-nitro-O-phenylenediamine
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: RAUWPNXIALNKQM-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1N)[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results