accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99-61-6

IUPAC Name: 3-nitrobenzaldehyde
Synonyms: 3-nitrobenzaldehyde M-nitrobenzaldehyde
Molecular Weight (g/mol): 151.12
Molecular Formula: C7H5NO3
InChi Key: ZETIVVHRRQLWFW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(C=O)=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)