accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99-62-7

IUPAC Name: 1,3-bis(propan-2-yl)benzene
Synonyms: 1,3-diisopropylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: UNEATYXSUBPPKP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(=CC=C1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results