accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99-77-4

IUPAC Name: ethyl 4-nitrobenzoate
Synonyms: ethyl-P-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: PHWSCBWNPZDYRI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results