accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 990-91-0

IUPAC Name: dibenzyl {[bis(benzyloxy)phosphoryl]oxy}phosphonate
Synonyms: dibenzyl (dibenzyloxyphosphoryl)oxyphosphonate
Molecular Weight (g/mol): 538.47
Molecular Formula: C28H28O7P2
InChi Key: NSBNXCZCLRBQTA-UHFFFAOYSA-N
SMILES: O=P(OCC1=CC=CC=C1)(OCC1=CC=CC=C1)OP(=O)(OCC1=CC=CC=C1)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)