accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 992-04-1

CAS RN 992-04-1

IUPAC Name: 2,3,4,5,6-pentaphenyl-1,1'-biphenyl
Synonyms: hexaphenylbenzene
Molecular Weight (g/mol): 534.7
Molecular Formula: C42H30
InChi Key: QBHWPVJPWQGYDS-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C(C2=CC=CC=C2)C(C2=CC=CC=C2)=C(C2=CC=CC=C2)C(C2=CC=CC=C2)=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results