accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3H3CH3CHFHFF

CAS RN 99337-56-1

IUPAC Name: tetrabutylazanium dihydrofluoride fluoride
Synonyms: tetrabutylammonium ion dihydrofluoride fluoride
Molecular Weight (g/mol): 301.48
Molecular Formula: C16H38F3N
InChi Key: MRXQMNWIADOAJY-UHFFFAOYSA-M
SMILES: F.F.[F-].CCCC[N+](CCCC)(CCCC)CCCC
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results