CAS RN 994-28-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
197.28
Molecular Formula:
C6H17ClGe
InChi Key:
FJAWDWVPBHGANJ-UHFFFAOYSA-N
SMILES:
Cl.CC[GeH](CC)CC
1
–
1
of
1
results
1
Triethylgermanium chloride, Thermo Scientific™
CAS: 994-28-5 Molecular Formula: C6H17ClGe Molecular Weight (g/mol): 197.28 MDL Number: MFCD00013591 InChI Key: FJAWDWVPBHGANJ-UHFFFAOYSA-N Synonym: triethylgermanium chloride,triethylchlorogermane,chloro triethyl germane,triethylchlorogermene,ccris 2650,triethylgermyl chloride,germanium triethyl chloride,acmc-209sco,chlorotriethylgermane PubChem CID: 70438 IUPAC Name: chloro(triethyl)germane SMILES: Cl.CC[GeH](CC)CC