accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClOFFFCl

CAS RN 99479-66-0

IUPAC Name: 2,6-dichloro-4-(trifluoromethoxy)aniline
Synonyms: 2,6-dichloro-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 246.01
Molecular Formula: C7H4Cl2F3NO
InChi Key: FKISQWQHZULEEG-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(OC(F)(F)F)C=C1Cl
Percent Purity 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)