accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99584-85-7

IUPAC Name: 3-methyl-4-nitrobenzamide
Synonyms: 3-methyl-4-nitrobenzamide
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: BCAAGQMIPKPDDX-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1[N+]([O-])=O)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results