accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99754-06-0

IUPAC Name: 4-chloro-2-[(2E)-3-(4-pentylphenyl)prop-2-enamido]benzoic acid
Synonyms: 4-chloro-2-[(2E)-3-(4-pentylphenyl)prop-2-enamido]benzoic acid
Molecular Weight (g/mol): 371.86
Molecular Formula: C21H22ClNO3
InChi Key: MDVFITMPFHDRBZ-JLHYYAGUSA-N
SMILES: CCCCCC1=CC=C(\C=C\C(=O)NC2=CC(Cl)=CC=C2C(O)=O)C=C1
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results