accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 998-30-1

CH3OSiOH3COCH3

CAS RN 998-30-1

IUPAC Name: triethoxysilyl
Synonyms: triethoxysilane
Molecular Weight (g/mol): 163.27
Molecular Formula: C6H15O3Si
InChi Key: PKDCQJMRWCHQOH-UHFFFAOYSA-N
SMILES: CCO[Si](OCC)OCC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)