accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNBr

CAS RN 99848-78-9

IUPAC Name: 5-bromo-1-methyl-2,3-dihydro-1H-indole
Synonyms: 5-bromo-1-methyl-2,3-dihydroindole
Molecular Weight (g/mol): 212.09
Molecular Formula: C9H10BrN
InChi Key: YIRSVRZXIXHOSN-UHFFFAOYSA-N
SMILES: CN1CCC2=C1C=CC(Br)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results