accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOClOCH3(E)

CAS RN 99854-17-8

IUPAC Name: (2E)-3-(2-chloro-3,4-dimethoxyphenyl)prop-2-enoate
Synonyms: (2E)-3-(2-chloro-3,4-dimethoxyphenyl)prop-2-enoate
Molecular Weight (g/mol): 241.65
Molecular Formula: C11H10ClO4
InChi Key: JIPTVEJKXDVECY-GQCTYLIASA-M
SMILES: COC1=CC=C(\C=C\C([O-])=O)C(Cl)=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results