accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOOH

CAS RN 99924-18-2

IUPAC Name: 5-phenyl-1,3-oxazole-4-carboxylic acid
Synonyms: 5-phenyl-1,3-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: RUKDIKJSGDVSIF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(OC=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results