CAS RN 1746-81-2
CAS RN 1746-81-2
IUPAC Name:
1-(4-chlorophenyl)-3-methoxy-3-methylurea
Synonyms:
monolinuron
Molecular Weight (g/mol):
214.65
Molecular Formula:
C9H11ClN2O2
InChi Key:
LKJPSUCKSLORMF-UHFFFAOYSA-N
SMILES:
CON(C)C(=O)NC1=CC=C(Cl)C=C1