accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 357166-30-4

NHNH(S)OOOOOHNNOH2NNaNa

CAS RN 357166-30-4

IUPAC Name: bis(2-{[4-(2-{2-amino-4-oxo-1H,4H,7H-pyrrolo[2,3-d]pyrimidin-5-yl}ethyl)phenyl]formamido}pentanedioic acid) pentahydrate tetrasodium
Synonyms: bis(pemetrexed) pentahydrate tetrasodium
Molecular Weight (g/mol): 1036.87
Molecular Formula: C40H52N10Na4O17
InChi Key: MNDQQGBMLGJNME-UHFFFAOYNA-N
SMILES: O.O.O.O.O.[Na].[Na].[Na].[Na].NC1=NC(=O)C2=C(NC=C2CCC2=CC=C(C=C2)C(=O)NC(CCC(O)=O)C(O)=O)N1.NC1=NC(=O)C2=C(NC=C2CCC2=CC=C(C=C2)C(=O)NC(CCC(O)=O)C(O)=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results