CAS RN 4253-89-8
CAS RN 4253-89-8
IUPAC Name:
2-(propan-2-yldisulfanyl)propane
Synonyms:
isopropyl disulfide
Molecular Weight (g/mol):
150.3
Molecular Formula:
C6H14S2
InChi Key:
LZAZXBXPKRULLB-UHFFFAOYSA-N
SMILES:
CC(C)SSC(C)C