accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63182-08-1

SOOOH2CNaCH2H2Ccomonmon

CAS RN 63182-08-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 338.4
Molecular Formula: C18H19NaO3S
InChi Key:
SMILES: [Na+].*-C=C.*-C=C.C=CC1=CC=CC=C1.[O-]S(=O)(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)