CAS RN 766485-15-8
CAS RN 766485-15-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-[2-[4-(6-Fluoro-1,2-benzoxazol-3-yl)-1-piperidyl]ethyl]-9-hydroxy-2-methyl-pyrido[1,2-a]pyrimidin-4-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.