accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7719-12-2

ClPClCl

CAS RN 7719-12-2

IUPAC Name: trichlorophosphane
Synonyms: phosphorus trichloride PCl3
Molecular Weight (g/mol): 137.32
Molecular Formula: Cl3P
InChi Key: FAIAAWCVCHQXDN-UHFFFAOYSA-N
SMILES: ClP(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results