missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-4-Hydroxypropranolol HCl, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | (+/-)-4-Hydroxy Propranolol Hydrochloride |
| CAS | 14133-90-5 |
| Molecular Formula | C16 H21 N O3 . Cl H |
| Quantity | 0.01 g |
| Synonym | 1-Naphthalenol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride (9 CI), 2-Propanol, 1-[(4-hydroxy-1-naphthyl)oxy]-3-(isopropylamino)-, hydrochloride (8 CI), 4-Hydroxypropranolol hydrochloride, 1-Naphthalenol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride (1:1), 4-Hydroxypropranolol Hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO3.ClH/c1-11(2)17-9-12(18)10-20-16-8-7-15(19)13-5-3-4-6-14(13)16;/h3-8,11-12,17-19 H,9-10H2,1-2H3;1 H |
| SMILES | Cl.CC(C)NCC(O)COc1ccc(O)c2ccccc12 |
| IUPAC Name | 4-[2-hydroxy-3-(propan-2-ylamino)propoxy]naphthalen-1-ol;hydrochloride |
| Molecular Weight (g/mol) | 311.8038 |
| Formula Weight | 311.1288 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction