Learn More
N-trans-Cinnamoylglycine, CDN
Product Code. 30326386
Click to view available options
Quantity:
0.025 g
Unit Size:
0.03g
Product Code. 30326386
Brand: CDN CDNX72900.025G
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Cinnamylglycine
CAS 16534-24-0
Molecular Formula C11 H11 N O3
Synonym Glycine, N-(1-oxo-3-phenyl-2-propen-1-yl)-, Glycine, N-(1-oxo-3-phenyl-2-propenyl)- (9 CI), Glycine, N-cinnamoyl- (6 CI,8 CI), N-(1-Oxo-3-phenyl-2-propen-1-yl)glycine, Cinnamoylglycine, N-Cinnamoylglycine, N-Cinnamylglycine, 2-Cinnamamidoacetic acid, 2-(3-Phenylacrylamido)acetic acid, 2-(3-Phenylprop-2-enamido)acetic acid, [(3-Phenylprop-2-enoyl)amino]acetic acid
InChI Formula InChI=1 S/C11H11NO3/c13-10(12-8-11(14)15)7-6-9-4-2-1-3-5-9/h1-7 H,8H2,(H,12,13)(H,14,15)
SMILES OC(=O)CNC(=O)C=Cc1ccccc1
IUPAC Name 2-(3-phenylprop-2-enoylamino)acetic acid
Molecular Weight (g/mol) 205.21
Quantity 0.025 g
Formula Weight 205.0739 g/mol
Percent Purity min 98% chemical purity
Shipping Condition Room Temperature