Learn More
Fisherbrand™ 0.2µm PES Capsule Filter
Provides additional final filtration. Fisher Scientific™ 0.2μm PES Capsule Filter is for use with Accu100 Ultrapure Water System.
Brand:  Fisherbrand™
Additional Details : Weight : 0.08460kg
Product Code. 15409659
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Constructed of PES
  • 0.2μm pore size
  • Removes final contaminants
  • For use with Accu100 Ultrapure Water System
Specifications
Specifications
Accu100 Ultrapure Water System
Capsule Filter
PES