Learn More
Trajan Scientific and Medical™ 0.36mm PEEKsil™ Tubing
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
Brand:  Trajan Scientific and Medical™ 0624374
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218793
  • 55.90€ / Pack of 2
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Specifications
Tubing
2/Pk.
0.025 mm
250 m